accessibility menu, dialog, popup

UserName

H3COOHNNOOCH3

CAS RN 381670-28-6

IUPAC Name: ethyl N-(2-methyl-3-nitrophenyl)carbamate
Synonyms: ethyl N-(2-methyl-3-nitrophenyl)carbamate
Molecular Weight (g/mol): 224.22
Molecular Formula: C10H12N2O4
InChi Key: UTVICKYWFUXPCS-UHFFFAOYSA-N
SMILES: CCOC(=O)NC1=C(C)C(=CC=C1)[N+]([O-])=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results