accessibility menu, dialog, popup

UserName

CH3NO

CAS RN 37928-17-9

IUPAC Name: 5-methyl-3,4-diphenyl-1,2-oxazole
Synonyms: 5-methyl-3,4-diphenyl-1,2-oxazole
Molecular Weight (g/mol): 235.29
Molecular Formula: C16H13NO
InChi Key: ZXIRUKJWLADSJS-UHFFFAOYSA-N
SMILES: CC1=C(C(=NO1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results