accessibility menu, dialog, popup

UserName

FF

CAS RN 37847-52-2

IUPAC Name: 2,4-difluoro-1,1'-biphenyl
Synonyms: 2,4-difluoro-1,1'-biphenyl
Molecular Weight (g/mol): 190.19
Molecular Formula: C12H8F2
InChi Key: JVHAJKHGPDDEEU-UHFFFAOYSA-N
SMILES: FC1=CC=C(C(F)=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results