accessibility menu, dialog, popup

UserName

CH3NFBr

CAS RN 375368-83-5

IUPAC Name: 3-bromo-6-fluoro-2-methylpyridine
Synonyms: 3-bromo-6-fluoro-2-methylpyridine
Molecular Weight (g/mol): 190.02
Molecular Formula: C6H5BrFN
InChi Key: GUYGQQWIKZLHTP-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=CC(F)=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results