accessibility menu, dialog, popup

UserName

BFFFFCH3NNOH

CAS RN 374564-83-7

IUPAC Name: 1-(2-hydroxyethyl)-3-methyl-1H-imidazol-3-ium; tetrafluoroboranuide
Synonyms: 3-(2-hydroxyethyl)-1-methylimidazol-1-ium tetrafluoroborate
Molecular Weight (g/mol): 213.97
Molecular Formula: C6H11BF4N2O
InChi Key: KLTUZFZUYLXTFF-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.C[N+]1=CN(CCO)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results