accessibility menu, dialog, popup

UserName

FFFFFFFI

CAS RN 374-98-1

IUPAC Name: 1,1,1,2,2,3,3-heptafluoro-4-iodobutane
Synonyms: 1,1,1,2,2,3,3-heptafluoro-4-iodobutane
Molecular Weight (g/mol): 309.95
Molecular Formula: C4H2F7I
InChi Key: AFHPVXVEXINDFS-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)CI
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)