accessibility menu, dialog, popup

UserName

CH3OHN(R)(R)OH(S)OH(R)HOOOOOCH3

CAS RN 37067-30-4

IUPAC Name: N-[(3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-[(4-methyl-2-oxo-2H-chromen-7-yl)oxy]oxan-3-yl]acetamide
Synonyms: N-[(3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-[(4-methyl-2-oxochromen-7-yl)oxy]oxan-3-yl]acetamide
Molecular Weight (g/mol): 379.37
Molecular Formula: C18H21NO8
InChi Key: QCTHLCFVVACBSA-XWEABGILSA-N
SMILES: CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)OC1OC1=CC=C2C(C)=CC(=O)OC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)