accessibility menu, dialog, popup

UserName

NH2SOOSH2NOO

CAS RN 3701-01-7

IUPAC Name: benzene-1,3-disulfonamide
Synonyms: 1,3-benzenedisulfonamide
Molecular Weight (g/mol): 236.26
Molecular Formula: C6H8N2O4S2
InChi Key: HUYYFHGIHVULSU-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC(=CC=C1)S(N)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results