accessibility menu, dialog, popup

UserName

CAS RN 36923-21-4

NSOH2NOO

CAS RN 36923-21-4

IUPAC Name: diphenylmethyl 7-amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Synonyms: diphenylmethyl 7-amino-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Molecular Weight (g/mol): 366.44
Molecular Formula: C20H18N2O3S
InChi Key: NYYBPASOTOAXQW-UHFFFAOYNA-N
SMILES: NC1C2SCC=C(N2C1=O)C(=O)OC(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results