CAS RN 367-83-9
IUPAC Name:
2-fluoro-5-methoxybenzoic acid
Synonyms:
2-fluoro-5-methoxybenzoic acid
Molecular Weight (g/mol):
170.14
Molecular Formula:
C8H7FO3
InChi Key:
REDSLTKMNCCQBC-UHFFFAOYSA-N
SMILES:
COC1=CC(C(O)=O)=C(F)C=C1
1
–
1
of
1
results
1
2-Fluoro-5-methoxybenzoic acid, 97+%
CAS: 367-83-9 Molecular Formula: C8H7FO3 Molecular Weight (g/mol): 170.139 MDL Number: MFCD00272566 InChI Key: REDSLTKMNCCQBC-UHFFFAOYSA-N PubChem CID: 2774543 IUPAC Name: 2-fluoro-5-methoxybenzoic acid SMILES: COC1=CC(=C(C=C1)F)C(=O)O