accessibility menu, dialog, popup

UserName

OOHNH2OHOHOHHO

CAS RN 3646-68-2

IUPAC Name: 2-amino-3,4,5,6-tetrahydroxyhexanoic acid
Synonyms: glucosaminic acid D-glucosamic acid glucosaminate D-glucosaminic acid
Molecular Weight (g/mol): 195.17
Molecular Formula: C6H13NO6
InChi Key: UFYKDFXCZBTLOO-UHFFFAOYNA-N
SMILES: NC(C(O)C(O)C(O)CO)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results