accessibility menu, dialog, popup

UserName

CAS RN 364-30-7

NH2FFNOO

CAS RN 364-30-7

IUPAC Name: 2,4-difluoro-6-nitroaniline
Synonyms: 2,4-difluoro-6-nitroaniline
Molecular Weight (g/mol): 174.11
Molecular Formula: C6H4F2N2O2
InChi Key: WPEUQAOOZXFRKZ-UHFFFAOYSA-N
SMILES: NC1=C(F)C=C(F)C=C1[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results