missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Difluoro-6-nitroaniline 98.0+%, TCI America™
Specifications
Specifications
| CAS | 364-30-7 |
| Melting Point | 84°C |
| Color | Yellow |
| Molecular Formula | C6H4F2N2O2 |
| Quantity | 5 g |
| MDL Number | MFCD00034058 |
| InChI Key | WPEUQAOOZXFRKZ-UHFFFAOYSA-N |
| SMILES | NC1=C(F)C=C(F)C=C1[N+]([O-])=O |
| IUPAC Name | 2,4-difluoro-6-nitroaniline |
| Molecular Weight (g/mol) | 174.11 |
| Show More |
Safety and Handling
| RTECSNumber | BX3730000 |
| TSCA | No |