CAS RN 36330-85-5
IUPAC Name:
4-{[1,1'-biphenyl]-4-yl}-4-oxobutanoic acid
Synonyms:
fenbufen cinopal wyeth brand of fenbufen fenbufen (product) lederfen fenbufenum 芬布芬 bufemid fenbufene fenbufen (substance)
Molecular Weight (g/mol):
254.29
Molecular Formula:
C16H14O3
InChi Key:
ZPAKPRAICRBAOD-UHFFFAOYSA-N
SMILES:
OC(=O)CCC(=O)C1=CC=C(C=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
4-(4-Biphenylyl)-4-oxobutyric acid, 96%
4-(4-Biphenylyl)-4-oxobutyric acid, 96%
CAS: 36330-85-5 Molecular Formula: C16H14O3 Molecular Weight (g/mol): 254.285 MDL Number: MFCD00056701 InChI Key: ZPAKPRAICRBAOD-UHFFFAOYSA-N Synonym: fenbufen,lederfen,cinopal,bufemid,napanol,4-4-biphenylyl-4-oxobutyric acid,3-4-phenylbenzoyl propionic acid,3-4-biphenylylcarbonyl propionic acid,4-biphenyl-4-yl-4-oxobutanoic acid,gamma-oxo 1,1'-biphenyl-4-butanoic acid PubChem CID: 3335 ChEBI: CHEBI:31599 IUPAC Name: 4-oxo-4-(4-phenylphenyl)butanoic acid SMILES: C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)CCC(=O)O