CAS RN 362661-83-4
IUPAC Name:
pyrazolo[1,5-a]pyridine-7-carbaldehyde
Synonyms:
pyrazolo[1,5-a]pyridine-7-carbaldehyde
Molecular Weight (g/mol):
146.15
Molecular Formula:
C8H6N2O
InChi Key:
SGNBVQKSJWCGOJ-UHFFFAOYSA-N
SMILES:
O=CC1=CC=CC2=CC=NN12
1
–
1
of
1
results
1
Promotions Available
Pyrazolo[1,5-a]pyridine-7-carboxaldehyde, 97%
Pyrazolo[1,5-a]pyridine-7-carboxaldehyde, 97%
CAS: 362661-83-4 Molecular Formula: C8H6N2O Molecular Weight (g/mol): 146.149 MDL Number: MFCD06739029 InChI Key: SGNBVQKSJWCGOJ-UHFFFAOYSA-N Synonym: pyrazolo 1,5-a pyridine-7-carbaldehyde,pyrazolo 1,5-a pyridine-7-carboxaldehyde,8-hydropyrazolo 1,5-a pyridine-7-carbaldehyde,pyrazolo 1,5-a pyridine-7-carboxaldehyde 9ci,pyrazolo 1,5-alpha pyridine-7-carboxaldehyde PubChem CID: 10866474 IUPAC Name: pyrazolo[1,5-a]pyridine-7-carbaldehyde SMILES: C1=CC2=CC=NN2C(=C1)C=O