accessibility menu, dialog, popup

UserName

CH3NONBr

CAS RN 362529-03-1

IUPAC Name: 5-[4-(bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole
Synonyms: 5-[4-(bromomethyl)phenyl]-3-methyl-1,2,4-oxadiazole
Molecular Weight (g/mol): 253.1
Molecular Formula: C10H9BrN2O
InChi Key: RWTXHMRWDVQEOV-UHFFFAOYSA-N
SMILES: CC1=NOC(=N1)C1=CC=C(CBr)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)