accessibility menu, dialog, popup

UserName

CAS RN 3615-41-6

H3COHOHOHOHO

CAS RN 3615-41-6

IUPAC Name: 2,3,4,5-tetrahydroxyhexanal
Synonyms: L(+)-rhamnose
Molecular Weight (g/mol): 164.16
Molecular Formula: C6H12O5
InChi Key: PNNNRSAQSRJVSB-UHFFFAOYNA-N
SMILES: CC(O)C(O)C(O)C(O)C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results