accessibility menu, dialog, popup

UserName

H2NOFFF

CAS RN 360-64-5

IUPAC Name: 2-(trifluoromethyl)benzamide
Synonyms: 2-trifluoromethylbenzamide
Molecular Weight (g/mol): 189.14
Molecular Formula: C8H6F3NO
InChi Key: QBAYIBZITZBSFO-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC=C1C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results