accessibility menu, dialog, popup

UserName

CAS RN 35873-49-5

CH3NONCH3NHNO

CAS RN 35873-49-5

IUPAC Name: 8-cyclopentyl-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: 8-cyclopentyl-1,3-dimethyl-7H-purine-2,6-dione
Molecular Weight (g/mol): 248.29
Molecular Formula: C12H16N4O2
InChi Key: SCVHFRLUNIOSGI-UHFFFAOYSA-N
SMILES: CN1C2=C(NC(=N2)C2CCCC2)C(=O)N(C)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results