missing translation for 'onlineSavingsMsg'
Learn More

8-Cyclopentyl-1,3-dimethylxanthine

Catalog No. AAJ61565MC
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61565MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61565MC Supplier Thermo Scientific Chemicals Supplier No. J61565MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A selective A1 adenosine receptor antagonist | CAS: 35873-49-5 | C12H16N4O2 | 248.286 g/mol

A selective A1 adenosine receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

CAS 35873-49-5
Melting Point 250°C to 252°C
Color White
Molecular Formula C12H16N4O2
Quantity 100 mg
MDL Number MFCD00055116
Synonym 8-cyclopentyltheophylline, 8-cyclopentyl-1,3-dimethylxanthine, 8-cyclopentyl theophylline, 1,3-dimethyl-8-cyclopentylxanthine, xanthine, 8-cyclopentyl-1,3-dimethyl, theophylline, 8-cyclopentyl, 8-cyclopentyl-1,3-dimethyl-1h-purine-2,6 3h,7h-dione, 1h-purine-2,6-dione, 8-cyclopentyl-3,9-dihydro-1,3-dimethyl, spectrum_000351, lopac-c-102
Solubility Information Soluble in 0;1M NaOH; Slightly soluble in water; Also soluble in DMSO at 16mg/ml at 60°C.
InChI Key SCVHFRLUNIOSGI-UHFFFAOYSA-N
SMILES CN1C2=C(C(=O)N(C1=O)C)NC(=N2)C3CCCC3
IUPAC Name 8-cyclopentyl-1,3-dimethyl-7H-purine-2,6-dione
Molecular Weight (g/mol) 248.286
PubChem CID 1917
Formula Weight 248.28
Physical Form Powder
Chemical Name or Material 8-Cyclopentyl-1,3-dimethylxanthine
Show More Show Less
RTECSNumber XH5093600
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only