accessibility menu, dialog, popup

UserName

CH3OOO

CAS RN 35817-27-7

IUPAC Name: 4-ethoxy-2H-chromen-2-one
Synonyms: 4-ethoxychromen-2-one
Molecular Weight (g/mol): 190.2
Molecular Formula: C11H10O3
InChi Key: FLZDKBHAUILYAZ-UHFFFAOYSA-N
SMILES: CCOC1=CC(=O)OC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results