accessibility menu, dialog, popup

UserName

CH3H3CCH3OOHNOOOOH

CAS RN 35793-73-8

IUPAC Name: 5-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoic acid
Synonyms: 5-(benzyloxy)-2-[(tert-butoxycarbonyl)amino]-5-oxopentanoic acid
Molecular Weight (g/mol): 337.37
Molecular Formula: C17H23NO6
InChi Key: AJDUMMXHVCMISJ-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCC(=O)OCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results