CAS RN 357611-51-9
CAS RN 357611-51-9
IUPAC Name:
(3,5-dimethyl-4-propoxyphenyl)boronic acid
Synonyms:
3,5-dimethyl-4-propoxyphenylboronic acid
Molecular Weight (g/mol):
208.06
Molecular Formula:
C11H17BO3
InChi Key:
ISUPZUFVQLUFLM-UHFFFAOYSA-N
SMILES:
CCCOC1=C(C)C=C(C=C1C)B(O)O