accessibility menu, dialog, popup

UserName

H3COSiOCH3OH3COCH3

CAS RN 35692-27-4

IUPAC Name: trimethoxy(4-methoxyphenyl)silane
Synonyms: trimethoxy(4-methoxyphenyl)silane
Molecular Weight (g/mol): 228.32
Molecular Formula: C10H16O4Si
InChi Key: ZESWBFKRPIRQCD-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)[Si](OC)(OC)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results