accessibility menu, dialog, popup

UserName

CH3CH3BrCH3CH3

CAS RN 355135-07-8

IUPAC Name: 2-bromo-9,9-bis(2-ethylhexyl)-9H-fluorene
Synonyms: 2-bromo-9,9-bis(2-ethylhexyl)fluorene
Molecular Weight (g/mol): 469.55
Molecular Formula: C29H41Br
InChi Key: JAWAIXIROBPNSH-UHFFFAOYNA-N
SMILES: CCCCC(CC)CC1(CC(CC)CCCC)C2=CC=CC=C2C2=C1C=C(Br)C=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results