accessibility menu, dialog, popup

UserName

H3COOClCl

CAS RN 35112-27-7

IUPAC Name: ethyl 2,5-dichlorobenzoate
Synonyms: ethyl 2,5-dichlorobenzoate
Molecular Weight (g/mol): 219.06
Molecular Formula: C9H8Cl2O2
InChi Key: JSZYWIKNIZKJAN-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC(Cl)=CC=C1Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results