accessibility menu, dialog, popup

UserName

HNOHOF

CAS RN 351-95-1

IUPAC Name: 2-[(4-fluorophenyl)amino]acetic acid
Synonyms: [(4-fluorophenyl)amino]acetic acid
Molecular Weight (g/mol): 169.16
Molecular Formula: C8H8FNO2
InChi Key: ZFIMCCCQJDZLCT-UHFFFAOYSA-N
SMILES: OC(=O)CNC1=CC=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results