accessibility menu, dialog, popup

UserName

CAS RN 34581-92-5

CH3ONOH3C

CAS RN 34581-92-5

IUPAC Name: 1-acetyl-4-methyl-2,5-dihydro-1H-pyrrol-2-one
Synonyms: 1-acetyl-4-methyl-5H-pyrrol-2-one
Molecular Weight (g/mol): 139.15
Molecular Formula: C7H9NO2
InChi Key: AINPDQRRTXHBAY-UHFFFAOYSA-N
SMILES: CC(=O)N1CC(C)=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results