missing translation for 'onlineSavingsMsg'
Learn More

1-Acetyl-4-methyl-2,5-dihydro-1H-pyrrol-2-one, 97%, Thermo Scientific™

Catalog No. BTB09313DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB09313DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB09313DA Supplier Thermo Scientific Chemicals Supplier No. BTB09313DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 34581-92-5
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H9NO2
Quantity 1 g
MDL Number MFCD00173863
Synonym 1-acetyl-4-methyl-2,5-dihydro-1h-pyrrol-2-one, 1-acetyl-4-methyl-5h-pyrrol-2-one, 1-acetyl-4-methyl-1h-pyrrol-2 5h-one, 2h-pyrrol-2-one,1-acetyl-1,5-dihydro-4-methyl
InChI Key AINPDQRRTXHBAY-UHFFFAOYSA-N
SMILES CC1=CC(=O)N(C1)C(=O)C
IUPAC Name 1-acetyl-3-methyl-2H-pyrrol-5-one
Molecular Weight (g/mol) 139.154
PubChem CID 2774997
Formula Weight 139
Percent Purity 97%
Chemical Name or Material 1-Acetyl-4-methyl-2, 5-dihydro-1H-pyrrol-2-one
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.