accessibility menu, dialog, popup

UserName

CH3OOOOCH3(E/Z)

CAS RN 34541-73-6

IUPAC Name: methyl 4-{2-[4-(methoxycarbonyl)phenyl]ethenyl}benzoate
Synonyms: methyl 4-{2-[4-(methoxycarbonyl)phenyl]ethenyl}benzoate
Molecular Weight (g/mol): 296.32
Molecular Formula: C18H16O4
InChi Key: JOODVYOWCWQPMV-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=CC2=CC=C(C=C2)C(=O)OC)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)