CAS RN 34513-98-9
IUPAC Name:
λ²-ruthenium(2+) iminooxidanium trinitrate
Synonyms:
λ²-ruthenium(2+) nitrosonium trinitrate
Molecular Weight (g/mol):
317.09
Molecular Formula:
N4O10Ru
InChi Key:
WOSOOWIGVAKGOC-UHFFFAOYSA-N
SMILES:
[Ru++].N#[O+].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
1
–
3
of
3
results
1
Ruthenium(III) nitrosylnitrate solution, Ru 1.5% w/v
CAS: 34513-98-9 MDL Number: MFCD00016313
2
Ruthenium(III) nitrosyl nitrate, in diluted aq. nitric acid solution
CAS: 34513-98-9 Molecular Formula: N4O10Ru Molecular Weight (g/mol): 317.09 MDL Number: MFCD00016313
3
Ruthenium(III) nitrosylnitrate, Ru 31.3% min
CAS: 34513-98-9 Molecular Formula: N4O10Ru MDL Number: MFCD00016313