CAS RN 3441-00-7
IUPAC Name:
5-(3-methylphenyl)-2H-1,2,3,4-tetrazole
Synonyms:
5-(3-methylphenyl)-2H-1,2,3,4-tetrazole
Molecular Weight (g/mol):
160.18
Molecular Formula:
C8H8N4
InChi Key:
GWZNHKMUSLKULV-UHFFFAOYSA-N
SMILES:
CC1=CC=CC(=C1)C1=NNN=N1
1
–
1
of
1
results
1
Promotions Available
5-(3-Methylphenyl)-1H-tetrazole, 99%
5-(3-Methylphenyl)-1H-tetrazole, 99%
CAS: 3441-00-7 Molecular Formula: C8H8N4 Molecular Weight (g/mol): 160.18 MDL Number: MFCD03093096 InChI Key: GWZNHKMUSLKULV-UHFFFAOYSA-N Synonym: 5-3-methylphenyl-1h-tetrazole,5-m-tolyl-1h-tetrazole,5-3-methylphenyl tetrazole,2h-tetrazole,5-3-methylphenyl,5-3-methylphenyl-1h-1,2,3,4-tetrazole,3-tolyl tetrazole,5-m-tolyl tetrazole,acmc-1cmun,5-m-tolyl 1h tetrazole PubChem CID: 583308 IUPAC Name: 5-(3-methylphenyl)-2H-tetrazole SMILES: CC1=CC=CC(=C1)C2=NNN=N2