accessibility menu, dialog, popup

UserName

CH3CH3SHHO

CAS RN 34300-94-2

IUPAC Name: 3-methyl-3-sulfanylbutan-1-ol
Synonyms: 3-mercapto-3-methylbutan-1-ol
Molecular Weight (g/mol): 120.21
Molecular Formula: C5H12OS
InChi Key: GBCGIJAYTBMFHI-UHFFFAOYSA-N
SMILES: CC(C)(S)CCO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results