accessibility menu, dialog, popup

UserName

I

CAS RN 34244-15-0

IUPAC Name: 1-iodopyrene
Synonyms: 1-iodopyrene
Molecular Weight (g/mol): 328.15
Molecular Formula: C16H9I
InChi Key: CWNJSSNWLUIMDP-UHFFFAOYSA-N
SMILES: IC1=C2C=CC3=CC=CC4=CC=C(C=C1)C2=C34
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results