accessibility menu, dialog, popup

UserName

H3COOOOOOCH3

CAS RN 33926-46-4

IUPAC Name: 4-ethoxyphenyl 4-{[(pentyloxy)carbonyl]oxy}benzoate
Synonyms: 4-ethoxyphenyl 4-{[(pentyloxy)carbonyl]oxy}benzoate
Molecular Weight (g/mol): 372.42
Molecular Formula: C21H24O6
InChi Key: QVUSHDHMLLZBDM-UHFFFAOYSA-N
SMILES: CCCCCOC(=O)OC1=CC=C(C=C1)C(=O)OC1=CC=C(OCC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results