accessibility menu, dialog, popup

UserName

H3COSOOCH3

CAS RN 3386-35-4

IUPAC Name: octyl 4-methylbenzene-1-sulfonate
Synonyms: octyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 284.41
Molecular Formula: C15H24O3S
InChi Key: LYQJBZLAANNIER-UHFFFAOYSA-N
SMILES: CCCCCCCCOS(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results