accessibility menu, dialog, popup

UserName

OHPOHHOO

CAS RN 33795-18-5

IUPAC Name: (4-hydroxyphenyl)phosphonic acid
Synonyms: 4-hydroxyphenylphosphonic acid
Molecular Weight (g/mol): 174.09
Molecular Formula: C6H7O4P
InChi Key: YIDVLWDHYNWHMH-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)P(O)(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results