accessibility menu, dialog, popup

UserName

CAS RN 33755-53-2

IUPAC Name: 4-nitrophenyl 5-[(3aS,4S,6aR)-2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanoate
Synonyms: 4-nitrophenyl 5-[(3aS,4S,6aR)-2-oxo-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoate
Molecular Weight (g/mol): 365.4
Molecular Formula: C16H19N3O5S
InChi Key: YUDNXDTXQPYKCA-YDHLFZDLSA-N
SMILES: [O-][N+](=O)C1=CC=C(OC(=O)CCCC[C@@H]2SC[C@@H]3NC(=O)N[C@H]23)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results