accessibility menu, dialog, popup

UserName

CAS RN 33685-54-0

IUPAC Name: tetrachloro(²H₂)ethane
Synonyms: tetrachloro(²H₂)ethane
Molecular Weight (g/mol): 169.85
Molecular Formula: C2H2Cl4
InChi Key: QPFMBZIOSGYJDE-QDNHWIQGSA-N
SMILES: [2H]C(Cl)(Cl)C([2H])(Cl)Cl
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results