accessibility menu, dialog, popup

UserName

CAS RN 3368-04-5

IUPAC Name: [(4-methyl-2-oxo-2H-chromen-7-yl)oxy]phosphonic acid
Synonyms: (4-methyl-2-oxochromen-7-yl)oxyphosphonic acid
Molecular Weight (g/mol): 256.15
Molecular Formula: C10H9O6P
InChi Key: BCHIXGBGRHLSBE-UHFFFAOYSA-N
SMILES: CC1=CC(=O)OC2=CC(OP(O)(O)=O)=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)