accessibility menu, dialog, popup

UserName

CH3FCl

CAS RN 33406-96-1

IUPAC Name: 1-chloro-4-fluoro-2-methylbenzene
Synonyms: 1-chloro-4-fluoro-2-methylbenzene
Molecular Weight (g/mol): 144.57
Molecular Formula: C7H6ClF
InChi Key: NIOGDCDTKPQEAT-UHFFFAOYSA-N
SMILES: CC1=CC(F)=CC=C1Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results