accessibility menu, dialog, popup

UserName

CAS RN 3317-61-1

IUPAC Name: 2,2-dimethyl-3,4-dihydro-2H-pyrrol-1-ium-1-olate
Synonyms: 2,2-dimethyl-3,4-dihydropyrrol-1-ium-1-olate
Molecular Weight (g/mol): 113.16
Molecular Formula: C6H11NO
InChi Key: VCUVETGKTILCLC-UHFFFAOYSA-N
SMILES: CC1(C)CCC=[N+]1[O-]
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results