accessibility menu, dialog, popup

UserName

H3COHOCH3CH3H3C

CAS RN 3302-10-1

IUPAC Name: 3,5,5-trimethylhexanoic acid
Synonyms: 3,5,5-trimethylhexanoic acid
Molecular Weight (g/mol): 158.24
Molecular Formula: C9H18O2
InChi Key: OILUAKBAMVLXGF-UHFFFAOYNA-N
SMILES: CC(CC(O)=O)CC(C)(C)C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)