accessibility menu, dialog, popup

UserName

CAS RN 32559-18-5

CH3OOHNHCl

CAS RN 32559-18-5

IUPAC Name: methyl piperidine-2-carboxylate hydrochloride
Synonyms: methyl piperidine-2-carboxylate hydrochloride
Molecular Weight (g/mol): 179.64
Molecular Formula: C7H14ClNO2
InChi Key: APCHKWZTSCBBJX-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C1CCCCN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results