accessibility menu, dialog, popup

UserName

H3CO

CAS RN 3234-28-4

IUPAC Name: 2-dodecyloxirane
Synonyms: 2-dodecyloxirane
Molecular Weight (g/mol): 212.38
Molecular Formula: C14H28O
InChi Key: IOHJQSFEAYDZGF-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCC1CO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results