accessibility menu, dialog, popup

UserName

H3CSiOCH3

CAS RN 321903-29-1

IUPAC Name: 1,1-dimethyl-1,3-dihydro-2,1-benzoxasilole
Synonyms: 1,1-dimethyl-3H-2,1-benzoxasilole
Molecular Weight (g/mol): 164.28
Molecular Formula: C9H12OSi
InChi Key: GLZOQCBCUKBUJG-UHFFFAOYSA-N
SMILES: C[Si]1(C)OCC2=CC=CC=C12
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results