accessibility menu, dialog, popup

UserName

F

CAS RN 321-38-0

IUPAC Name: 1-fluoronaphthalene
Synonyms: 1-fluoronaphthalene
Molecular Weight (g/mol): 146.16
Molecular Formula: C10H7F
InChi Key: CWLKTJOTWITYSI-UHFFFAOYSA-N
SMILES: FC1=C2C=CC=CC2=CC=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)