accessibility menu, dialog, popup

UserName

CAS RN 317-46-4

IUPAC Name: 2-fluoro-3-nitrobenzoic acid
Synonyms: 2-fluoro-3-nitrobenzoic acid
Molecular Weight (g/mol): 185.11
Molecular Formula: C7H4FNO4
InChi Key: WLGUSLGYTNJJFV-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C(=CC=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results