CAS RN 313981-55-4
IUPAC Name:
2,4,6-trimethyl-N-[2-(trifluoromethyl)phenyl]benzene-1-sulfonamide
Synonyms:
2,4,6-trimethyl-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide
Molecular Weight (g/mol):
343.36
Molecular Formula:
C16H16F3NO2S
InChi Key:
SKJJIFRWCCSXGL-UHFFFAOYSA-N
SMILES:
CC1=CC(C)=C(C(C)=C1)S(=O)(=O)NC1=CC=CC=C1C(F)(F)F
1
–
1
of
1
results
1
2,4,6-Trimethyl-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide, Thermo Scientific Chemicals
CAS: 313981-55-4 Molecular Formula: C16H16F3NO2S Molecular Weight (g/mol): 343.36 MDL Number: MFCD00784362 InChI Key: SKJJIFRWCCSXGL-UHFFFAOYSA-N Synonym: o-3m3fbs,2,4,6-trimethyl-n-2-trifluoromethyl phenyl benzenesulfonamide,tocris-1942,benzenesulfonamide,2,4,6-trimethyl-n-2-trifluoromethyl phenyl PubChem CID: 770820 IUPAC Name: 2,4,6-trimethyl-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide SMILES: CC1=CC(C)=C(C(C)=C1)S(=O)(=O)NC1=CC=CC=C1C(F)(F)F