accessibility menu, dialog, popup

UserName

BClFHOOH

CAS RN 313545-32-3

IUPAC Name: (2-chloro-6-fluorophenyl)boronic acid
Synonyms: 2-chloro-6-fluorophenylboronic acid
Molecular Weight (g/mol): 174.36
Molecular Formula: C6H5BClFO2
InChi Key: NXSZSZJWZVLHAY-UHFFFAOYSA-N
SMILES: OB(O)C1=C(F)C=CC=C1Cl
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results